Learn More
Trajan™ Diamond MS CTC PAL Autosampler Syringes
SDP
Catalog No. SG004770 Shop All SGE Inc. Products
Change view
Click to view available options
Volume (Metric):
25 μL
50 μL
Model:
25R-CTC-GT-MS1
50R-CTC-GT-MS1
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric) Model
SG004770 50 μL 50R-CTC-GT-MS1
SG003770 25 μL 25R-CTC-GT-MS1
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. SG004770 Supplier Trajan™ Supplier No. 004770
Improves the sensitivity of your MS analysis with reduced non specific sample interaction and increased analyte recovery.
  • Lowest syringe carryover
  • Reduced non specific sample interaction and increased analyte recovery
  • Ultra smooth removable needles with choice of surface treatment.
Specifications
For Use With (Application) Gas/Liquid chromatography
Product Type Diamond MS Syringe
Max. Temperature (English) 662°F
Max. Temperature (Metric) 350°C
Model 50R-CTC-GT-MS1
Quantity Ea.
Volume (Metric) 50 μL
For Use With (Equipment) CTC PAL Autosampler Syringe
Connection Type Removable