Description
- Lowest syringe carryover
- Reduced non specific sample interaction and increased analyte recovery
- Ultra smooth removable needles with choice of surface treatment.
Specifications
Specifications
| Includes | Removable ultra smooth needle |
| Product Type | Diamond MS Manual Syringe |
| Model | 500R-GT-MS1 |
| Quantity | Ea. |
| Volume (Metric) | 500 μL |
| Connection Type | Removable |