Learn More
Trajan™ Diamond MS Shimadzu Autosampler Syringes
SDP
Catalog No. SG002780 Shop All SGE Inc. Products
Change view
Click to view available options
Volume (Metric):
10 μL
Catalog No. Volume (Metric)
SG002780 10 μL
1 options
Catalog No. SG002780 Supplier Trajan™ Supplier No. 002780
Improves the sensitivity of your MS analysis with reduced non specific sample interaction and increased analyte recovery.
  • High quality solutions for liquid handling
  • Accurate and reproducible injections
  • Specifically designed for Shimadzu instruments
Specifications
Product Type Diamond MS Autosampler Syringe
Model 10R-S-GT-MS1
Quantity Ea.
Volume (Metric) 10 μL
Connection Type Removable