Learn More
Trajan™ Diamond MS Thermo RSH and CTC RTC Autosampler Syringes
SDP
Catalog No. SG004790 Shop All SGE Inc. Products
Change view
Click to view available options
Volume (Metric):
25 μL
50 μL
Model:
25R-RTC/RSH-GT-MS1
50R-RTC/RSH-GT-MS1
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric) Model
SG004790 50 μL 50R-RTC/RSH-GT-MS1
SG003790 25 μL 25R-RTC/RSH-GT-MS1
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. SG004790 Supplier Trajan™ Supplier No. 004790
Improves the sensitivity of your MS analysis with reduced non-specific sample interaction and increased analyte recovery.
  • Lowest syringe carryover
  • Reduced non-specific sample interaction and increased analyte recovery
  • Compatible with ultra smooth removable needles with choice of surface treatment (sold separately)
Specifications
For Use With (Application) Gas/Liquid chromatography
Product Type Autosampler Syringe
Max. Temperature (English) 662°F
Max. Temperature (Metric) 350°C
Model 50R-RTC/RSH-GT-MS1
Quantity Ea.
Volume (Metric) 50 μL
For Use With (Equipment) Thermo RSH and CTC RTC
Connection Type Removable