Learn More
Trajan™ Diamond MS Thermo RSH and CTC RTC Autosampler Syringes
SDP
Catalog No. SG005790 Shop All SGE Inc. Products
Change view
Click to view available options
Volume (Metric):
25 μL
50 μL
100 μL
Model:
100R-RTC/RSH-GT-MS1
25R-RTC/RSH-GT-MS1
50R-RTC/RSH-GT-MS1
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric) Model
SG005790 100 μL 100R-RTC/RSH-GT-MS1
SG003790 25 μL 25R-RTC/RSH-GT-MS1
SG004790 50 μL 50R-RTC/RSH-GT-MS1
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. SG005790 Supplier Trajan™ Supplier No. 005790
Improves the sensitivity of your MS analysis with reduced non-specific sample interaction and increased analyte recovery.
  • Lowest syringe carryover
  • Reduced non-specific sample interaction and increased analyte recovery
  • Compatible with ultra smooth removable needles with choice of surface treatment (sold separately)
Specifications
For Use With (Application) Gas/Liquid chromatography
Product Type Autosampler Syringe
Max. Temperature (English) 662°F
Max. Temperature (Metric) 350°C
Model 100R-RTC/RSH-GT-MS1
Quantity Ea.
Volume (Metric) 100 μL
For Use With (Equipment) Thermo RSH and CTC RTC
Connection Type Removable