Description
- Stainless steel 10-32 thread
- Inert contact surface makes it biocompatible
- Ideal for use with corrosive solvents
Specifications
Specifications
| Material | Stainless Steel |
| Type | Replacement PEEK Hexnut Ferrule |
| Max. Pressure | 5000 psi |
| Thread Size | 10-32 |
| Quantity | 10/Pk. |
| Includes | Pk. size: 10 |