Learn More
Trajan™ eVol™ MEPS™ Syringes
SDP
Catalog No. 03350321 Shop All SGE Inc. Products
Change view
Click to view available options
Product Type:
eVol™ Syringe
Catalog No. Product Type
03-350-321 eVol™ Syringe
1 options
Catalog No. 03-350-321 Supplier Trajan™ Supplier No. 2910028
For use with eVol™ Digital Analytical Syringe digitally controlled positive displacement dispensing system.
  • Enables MEPS (micro SPE) to be semi-automated and proofed before transition to fully automated platforms
  • 2011 Firmware required
  • Maximum operating temperature of 350°C
Specifications
For Use With (Application) Liquid handling procedure
Product Type eVol™ Syringe
For Use With (Equipment) MEPS BINs