Learn More
Trajan™ Hexnut Fittings
SDP
Catalog No. SG1021011 Shop All SGE Inc. Products
Change view
Click to view available options
Includes:
10 hexnuts and 10 ferrules
2 hexnuts and 2 ferrules
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Includes
SG-1021011 10 hexnuts and 10 ferrules
SG-1021003 2 hexnuts and 2 ferrules
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. SG-1021011 Supplier Trajan™ Supplier No. 1021011
HexNut fittings provide ideal connections in HPLC systems
  • Stainless steel 10-32 thread
  • Inert contact surface makes it biocompatible
  • Ideal for use with corrosive solvents
Specifications
Includes 10 hexnuts and 10 ferrules
Thread Size 10-32
Material Stainless Steel
Pressure 5000 psi