Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Trajan™ BP21 (FFAP) GC Capillary Columns
SDP

Catalog No. SG054468 Shop All SGE Inc. Products
Change view
Click to view available options
Length (Metric):
25 m
30 m
50 m
60 m
Diameter (Metric) Inner:
0.22 mm
0.25 mm
0.32 mm
0.53 mm
Film Thickness:
0.25 μm
0.5 μm
1 μm
10 product options available for selection
Product selection table with 10 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Length (Metric) Diameter (Metric) Inner Film Thickness
SG-054468 25 m 0.32 mm 0.25 μm
SG-054462 25 m 0.22 mm 0.25 μm
SG-054469 50 m 0.32 mm 0.25 μm
SG-054474 25 m 0.53 mm 0.5 μm
SG-054465 30 m 0.25 mm 0.25 μm
SG-054466 60 m 0.25 mm 0.25 μm
SG-054471 30 m 0.32 mm 0.25 μm
SG-054472 60 m 0.32 mm 0.25 μm
SG-054477 30 m 0.53 mm 0.5 μm
SG-054478 30 m 0.53 mm 1 μm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 10 options available.
10 options
Catalog No. SG-054468 Supplier Trajan™ Supplier No. 054468

A very polar nitroterephthalic acid modified PEG phase suited to the analysis of free fatty acids

  • Nitroterephthalic acid modified PEG polar phase
  • Ideal for low molecular weight acids
  • Applications include volatile free acids, fatty acid methyl esters (FAMEs), alcohols, aldehydes and ketones

Specifications

Diameter (Metric) Inner 0.32 mm
Stationary Phase Polyethylene Glycol
Functionality/Interaction Type Volatile Free Acids, Fatty Acid Methyl Esters, Alcohols, Aldehydes, Acrylates, Ketones
USP Type G25, G35
For Use With (Application) Acrylates Analysis, Fatty Acid Methyl Esters, Alcohol Analysis, Aldehydes Analysis, Acrylates, Ketones
Particle Size 0.25 μm
Temperature Range 35°C to 240/250°C
Film Thickness 0.25 μm
Length (Metric) 25 m
Packing Material BP21 (FFAP)
Polarity Polar
Coating Fused Silica
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.