Learn More
Trajan™ Gas and Vapor Sampler Kit
SDP
Catalog No. SG090111 Shop All SGE Inc. Products
Change view
Click to view available options
For gas and vapor collection
Catalog No.
SG-090111 For gas and vapor collection
1 options
Catalog No. SG-090111 Supplier Trajan™ Supplier No. 090111
Enables spot collection of environmental gas samples or low pressure gas streams in 200cm3 impervious resealable bags
  • Gas and vapor collection
  • Resealable sample bags
  • Ideal for field analysis
Specifications
Type Gas and Vapor Sampling Kit
Includes Canister, Bulb, 10 bags, Marker Pen and Instructions