Learn More
SGE Gastight™ Syringe, Fixed Needle (LTN), Sold by MilliporeSigma™ Supelco™
Catalog No. 111011649 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Volume (Metric):
10 μL
25 μL
50 μL
100 μL
250 μL
500 μL
1000 μL
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric)
11-101-1649 10 μL
11-101-1352 25 μL
11-100-4972 50 μL
11-101-1208 100 μL
11-101-6044 250 μL
11-101-1235 500 μL
11-101-1052 1000 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Catalog No. 11-101-1649 Supplier MilliporeSigma Supelco Supplier No. 26256
Add to Cart
Add to Cart
Suitable for manual dispenser/diluter applications. Maximum inertness for liquid or gas samples.
TRUSTED_SUSTAINABILITY
Specifications
Material Borosilicate Glass (Barrel), PTFE (Plunger), Stainless Steel (Tip)
Needle Gauge 26 G
Diameter (Metric) Outer Needle 0.47 mm
Model 10 F-GT
Needle Point Number 2
Quantity 1 Ea.
Volume (Metric) 10 μL
Length (Metric) Needle 50 mm
Needle Point Style Bevel
Connection Type Kel-F Luer Cones, Fixed
GASTIGHT is a registered trademark of Hamilton Co.