Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Trajan™ GC Capillary Column Connectors: Replacement μ-Unions
SDP

Catalog No. SG073574 Shop All SGE Inc. Products
Change view
Click to view available options
Connections:
0.1 - 0.32 mm and 0.53 mm
0.53 mm and 0.53 mm
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Connections
SG073574 0.53 mm and 0.53 mm
SG073573 0.1 - 0.32 mm and 0.53 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. SG073574 Supplier Trajan™ Supplier No. 073574

missing translation for 'validMainframeMsg'

SilTite μ-Union delivers an easy, leak-free installation for capillary columns without the use of any tools

  • Next generation ferrule system for gas chromatography
  • Eliminates leaks
  • No contamination
  • No need to retighten after temperature cycling
  • Cleaner spectra for better library matches

Specifications

Type Replacement μ-Union
Connections 0.53 mm and 0.53 mm
For Use With (Equipment) GC Capillary Column Connectors
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.