Learn More
Trajan™ Glass Lined Bulkhead Unions with Locknut
SDP
Catalog No. p-4824122 Shop All SGE Inc. Products
Change view
Click to view available options
Diameter (Metric) Inner:
0.4 mm
0.8 mm
Description:
0.25 mm restriction (at one end)
No restriction
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Diameter (Metric) Inner Description
SG-1236306 0.8 mm No restriction
SG-1236302 0.4 mm 0.25 mm restriction (at one end)
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. SG-1236306 Supplier Trajan™ Supplier No. 1236306
Glass lined stainless steel construcion. Bulkhead unions with locknut
  • Allows plate mounting of union
  • For use with fused silica
  • Low dead volume
  • High temperature applications
Specifications
Type Bulkhead Union
Description No restriction
Material Glass-lined Stainless Steel
Diameter (Metric) Inner 0.8 mm