Learn More
SGE™ Glass Lined Tubing Tee
SDP
Catalog No. SG0828010 Shop All SGE Inc. Products
Change view
Click to view available options
Diameter (Metric) Inner:
1 mm
Catalog No. Diameter (Metric) Inner
SG-0828010 1 mm
1 options
Catalog No. SG-0828010 Supplier SGE™ Supplier No. 0828010
GLTT/8 GLT Tee Tube Fittings
GLT exhibits excellent resistance to the harshest of conditions. Maximum temperature for continuous use of GLT is 500°C.
Specifications
Material GLT
Diameter (Metric) Inner 1 mm