Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SGE™ GLT™ Tubing
SDP

Catalog No. SG0827375 Shop All SGE Inc. Products
Change view
Click to view available options
Diameter (English) Inner:
0.02 in.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Diameter (English) Inner
SG-0827375 0.02 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. SG-0827375 Supplier SGE™ Supplier No. 0827375

missing translation for 'validMainframeMsg'

Specifications

For Use With HPLC column; GC/MS interface; FTIR transfer line; inlet liner; thermal desorption tube; tee piece; union; SFC
Diameter (English) Inner 0.02 in.
Diameter (Metric) Inner 0.5 mm
Diameter (Metric) Outer 3.2 mm
Overall Length (Metric) 30 cm
Material Glass Lined
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.