Learn More
Description
The MEPS approach to sample preparation is suitable for reversed phases, normal phases, mixed mode and ion exchange chemistries.
- For GC applications: needle is 23 gauge, 0.63mm OD, cone point style
- For LC applications: needle is 22 gauge, 0.72mm OD, LC point style
- Typical BIN life is conservatively about 40-100 samples but actual life is dependant on the sample matrix
- The dimensions of the sorbent bed ensure that the sorbent performance remains identical to conventional SPE devices used for extraction of similar samples
- Sample preparation for complex biological samples is readily adapted to MEPS and reduces the volume of sample and reagents required for extraction when compared with conventional SPE and other “micro extraction” procedures
- Needles are packed in a sealed blister pack of 5 needles to ensure their integrity
- Suitable for use with eVol™ automated syringes
Specifications
Specifications
| Material | Silica |
| Needle Gauge | 23 g |
| Diameter (English) Outer Needle | 0.024 in. |
| Diameter (Metric) Outer Needle | 0.06 cm |
| For Use With (Application) | MEPS |
| Product Type | MEPS Barrel Insert and Needle (BIN) |
| Needle Point Style | Cone |
| For Use With (Equipment) | HTA 300 A Plus, Thermo Scientific, Varian 8400 and 100 μL CTC Analytics System |
| Pore Size | 60 Å |
| Particle Size | 45 μm |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
