Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SGE™ MEPS™ BINs
SDP

Catalog No. SG2900409 Shop All SGE Inc. Products
Change view
Click to view available options
For Use With (Equipment):
250 μL CTC Analytics System
A300 Plus, Thermo Scientific, Varian 8400, 100 μL CTC Analytics Systems
HTA 300 A Plus, Thermo Scientific, Varian 8400 and 100 μL CTC Analytics System
Product Type:
MEPS Barrel Insert and Needle (BIN)
MEPS Development Kit
Sorbent:
C18
One each C18, C8, C2, Silica, C8+SCX
SAX
SCX
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment) Product Type Sorbent
SG2900409 A300 Plus, Thermo Scientific, Varian 8400, 100 μL CTC Analytics Systems MEPS Barrel Insert and Needle (BIN) SAX
03-350-136 HTA 300 A Plus, Thermo Scientific, Varian 8400 and 100 μL CTC Analytics System MEPS Barrel Insert and Needle (BIN) -
03-350-135 HTA 300 A Plus, Thermo Scientific, Varian 8400 and 100 μL CTC Analytics System MEPS Barrel Insert and Needle (BIN) C18
03-350-139 HTA 300 A Plus, Thermo Scientific, Varian 8400 and 100 μL CTC Analytics System MEPS Development Kit One each C18, C8, C2, Silica, C8+SCX
03-350-271 250 μL CTC Analytics System MEPS Barrel Insert and Needle (BIN) SCX
03-350-272 250 μL CTC Analytics System MEPS Barrel Insert and Needle (BIN) SAX
SG2900408 A300 Plus, Thermo Scientific, Varian 8400, 100 μL CTC Analytics Systems MEPS Barrel Insert and Needle (BIN) SCX
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Catalog No. SG2900409 Supplier SGE™ Supplier No. 2900409

Available in needle styles to suit LC and GC applications

The MEPS approach to sample preparation is suitable for reversed phases, normal phases, mixed mode and ion exchange chemistries.

  • For GC applications: needle is 23 gauge, 0.63mm OD, cone point style
  • For LC applications: needle is 22 gauge, 0.72mm OD, LC point style
  • Typical BIN life is conservatively about 40-100 samples but actual life is dependant on the sample matrix
  • The dimensions of the sorbent bed ensure that the sorbent performance remains identical to conventional SPE devices used for extraction of similar samples
  • Sample preparation for complex biological samples is readily adapted to MEPS and reduces the volume of sample and reagents required for extraction when compared with conventional SPE and other “micro extraction” procedures
  • Needles are packed in a sealed blister pack of 5 needles to ensure their integrity
  • Suitable for use with eVol™ automated syringes

Specifications

Material Silica
Needle Gauge 22 g
Diameter (English) Outer Needle 0.028 in.
Diameter (Metric) Outer Needle 0.07 cm
For Use With (Application) MEPS
Product Type MEPS Barrel Insert and Needle (BIN)
Needle Point Style LC
For Use With (Equipment) A300 Plus, Thermo Scientific, Varian 8400, 100 μL CTC Analytics Systems
Pore Size 60 Å
Particle Size 45 μm
Sorbent SAX
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.