Learn More
Trajan™ MEPS BINs for LC Applications
SDP
MEPS is Micro Extraction by Packed Sorbent. Provides miniaturization of conventional sample preparation devices
Supplier:  Trajan™ 2900403
 View more versions of this product
Catalog No. 03-350-149
Description
Description
  • Save hours in sample preparation
  • Reduce solvent consumption and sample volume
  • Available in reversed phase, normal phase, mixed mode and ion exchange chemistries
Specifications
Specifications
LC
HTA 300 APlus, Thermo Scientific, Varian 8400 and 100 μL CTC Analytics Systems
C8+SCX
10 μL to 1 mL or more
60 Å
0.72 mm
Liquid Chromatography
45 μm
MEPS Barrel Insert and Needle (BIN)
22 g
LC point needle
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.