Learn More
SGE™ MEPS™ Syringes
SDP
Catalog No. 03350134 Shop All SGE Inc. Products
Change view
Click to view available options
Volume (Metric):
100 μL
250 μL
For Use With (Equipment):
CTC Analytics
CTC Analytics, HTA 300APlus, Thermo Scientific and Varian 8400 System
HTA 300APlus, Thermo Scientific and Varian 8400 System
Product Type:
MEPS™ Syringe
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric) For Use With (Equipment) Product Type
03-350-134 250 μL CTC Analytics MEPS™ Syringe
03-350-133 250 μL HTA 300APlus, Thermo Scientific and Varian 8400 System MEPS™ Syringe
03-350-132 100 μL CTC Analytics, HTA 300APlus, Thermo Scientific and Varian 8400 System MEPS™ Syringe
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 03-350-134 Supplier SGE™ Supplier No. 006292
For use in micro-SPE
  • The eVol™ custom programming function enables MEPS to be semi-automated and proofed before transition to fully automated platforms
Specifications
Diameter (English) Inner 0.3 in.
Diameter (Metric) Inner 7.8 mm
For Use With (Application) Gas/Liquid chromatography
Product Type MEPS™ Syringe
Quantity Ea.
Volume (Metric) 250 μL
For Use With (Equipment) CTC Analytics
Connection Type Removable