Learn More
MilliporeSigma™ Supelco™ SGE MicroVolume Syringes, Standard Plunger
Catalog No. 111011116 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Volume (Metric):
25 μL
500 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric)
11-101-1116 500 μL
11-101-1648 25 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 11-101-1116 Supplier MilliporeSigma™ Supelco™ Supplier No. 26252
Add to Cart
Add to Cart
Equipped with bevel tip needles, designed for optimum septum penetration and to prevent septum coring
  • Sharp and clear fused graduation
  • Extended graduation lines and wide white backing for reduced errors
  • Angled needle tip for optimum septum penetration
  • Standard, stainless steel plunger is individually fitted to its barrel
  • Extended barrel guides the plunger during injection
  • Suitable for training and student use
  • Robust
TRUSTED_SUSTAINABILITY
Specifications
Material Stainless Steel (Tip)
Description Standard Plunger
Needle Gauge 25 G
Product Type SGE Microvolume Syringe
Model 500 F
Needle Point Number 2
Quantity 1 Ea.
Volume (Metric) 500 μL
Length (Metric) Needle 50 mm
Needle Point Style Bevel
Connection Type Fixed