Learn More
Trajan™ Mini Union Nut with 1.2 mm Hole
SDP
Catalog No. SG103400 Shop All SGE Inc. Products
Change view
Click to view available options
Quantity:
5/Pk.
Catalog No. Quantity
SG-103400 5/Pk.
1 options
Catalog No. SG-103400 Supplier Trajan™ Supplier No. 103400
Mini Union nut forms a soft grip to the capillary column for better reliability and ease of work. Delivers an easy, leak-free installation for capillary columns without the use of any tools
  • Easy to use
  • Forms a reliable seal
  • Reusable
  • Soft material
  • Forms a soft grip to the capillary column
  • Low emissions
Specifications
Type Nut
Quantity 5/Pk.
Material Stainless Steel