Learn More
MilliporeSigma™ Supelco™ SGE Needles for Luer Lock Syringes
Catalog No. 111011224 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Needle Point Style:
Bevel
Cone with Side Port
Length (Metric) Needle:
50 mm
70 mm
115 mm
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Needle Point Style Length (Metric) Needle
11-101-1224 Bevel 115 mm
11-101-1529 Bevel 50 mm
11-101-1303 Cone with Side Port 50 mm
11-101-1223 Bevel 70 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 11-101-1224 Supplier MilliporeSigma™ Supelco™ Supplier No. 26273U
Add to Cart
Add to Cart
Preferred for manual injection where piercing the septum in exactly the same place is difficult. The beveled tip is designed for optimum septum penetration and prevention of septum coring.
TRUSTED_SUSTAINABILITY
Specifications
Material Stainless Steel (Tip)
Description NLL-11.5/23
Needle Gauge 23 G
Diameter (Metric) Outer Needle 0.63 mm
Product Type SGE Needles
Needle Point Number 2
Quantity 5 Pk.
Length (Metric) Needle 115 mm
Needle Point Style Bevel
Connection Type Luer Lock