Learn More
Trajan™ Replacement Plungers
SDP
Catalog No. SG0318928 Shop All SGE Inc. Products
Change view
Click to view available options
For Use With (Equipment):
10 μL Removable Guided Plunger Syringe
10 μL Removable Needle Gas-tight Syringe for CTC/Carlo Erba/Fisons Autosampler
100 μL Fixed Needle Gas-tight Syringe for Carlo Erba, CTC, Fisons Autosamplers
Fixed Needle Gas Tight Syringe (10μL)
Syringe Models SG-003988, SG-003985, 25 μL
Syringe Needles (25μL), CTC Gas Tight Syringe
Syringe Needles (250μL), Gas Tight Syringe
Syringe Needles (50μL), Fixed Needle Gas Tight Syringe
Syringe Needles (500μL), CTC Gas Tight Syringe
Product Type:
Replacement Plunger, 10R-C/T-GT
Replacement Plunger, P10-GP-GT
Replacement Plunger, P100F-C/T-GT
Replacement Plunger, P10F-BT-GT
Replacement Plunger, P250R-C/T-GT
Replacement Plunger, P25F-CTC-GT
Replacement Plunger, P25R-C/T-GT
Replacement Plunger, P500F-CTC-GT
Replacement Plunger, P50F-C/T-GT
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment) Product Type
SG0318928 Syringe Needles (500μL), CTC Gas Tight Syringe Replacement Plunger, P500F-CTC-GT
SG-031805 10 μL Removable Guided Plunger Syringe Replacement Plunger, P10-GP-GT
SG0318922 Syringe Needles (25μL), CTC Gas Tight Syringe Replacement Plunger, P25F-CTC-GT
03-350-106 Syringe Models SG-003988, SG-003985, 25 μL Replacement Plunger, P25R-C/T-GT
03-350-110 100 μL Fixed Needle Gas-tight Syringe for Carlo Erba, CTC, Fisons Autosamplers Replacement Plunger, P100F-C/T-GT
03-350-105 10 μL Removable Needle Gas-tight Syringe for CTC/Carlo Erba/Fisons Autosampler Replacement Plunger, 10R-C/T-GT
03-350-108 Syringe Needles (50μL), Fixed Needle Gas Tight Syringe Replacement Plunger, P50F-C/T-GT
03-350-111 Syringe Needles (250μL), Gas Tight Syringe Replacement Plunger, P250R-C/T-GT
03-351-123 Fixed Needle Gas Tight Syringe (10μL) Replacement Plunger, P10F-BT-GT
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
Catalog No. SG0318928 Supplier Trajan™ Supplier No. 0318928
Replacement Plungers are available for all SGE gas tight syringes.
  • For use with SGE gas tight syringes.
  • Use the SGE Syringe Part Number to find a replacement plunger.
Specifications
Material PTFE (Tip)
For Use With (Application) IN VITRO use only. For lab analyzers only. Not for human use.
Product Type Replacement Plunger, P500F-CTC-GT
For Use With (Equipment) Syringe Needles (500μL), CTC Gas Tight Syringe