Description
- Next generation ferrule system for gas chromatography
- Eliminates leaks
- No contamination
- No need to retighten after temperature cycling
- Cleaner spectra for better library matches
Specifications
Specifications
| Material | SilTite Metal |
| For Use With (Equipment) | Shimadzu Injectors and Non-MS Detectors |
| Type | GC Ferrule and Nut Kit |
| Quantity | 10 ferrules, 2 nuts |
| Diameter (Metric) Inside | 0.32 mm |