Learn More
SGE™ SilTite™ Ferrule Starter Kits Replacement SilTite Nuts and Base Seals
SDP
Catalog No. SG073226 Shop All SGE Inc. Products
Change view
Click to view available options
Type:
Inlet Gold Base Seal
Nut
For Use With (Equipment):
Agilent Split/Splitless Injector
Split/Splitless Injectors
Quantity:
10/Pk.
2/Pk.
5/Pk.
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Type For Use With (Equipment) Quantity
SG073226 Nut Split/Splitless Injectors 5/Pk.
SG073400 Inlet Gold Base Seal Agilent Split/Splitless Injector 2/Pk.
SG073401 Inlet Gold Base Seal Agilent Split/Splitless Injector 10/Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. SG073226 Supplier SGE™ Supplier No. 073226
Specially designed ferrule and nut system developed specifically for fused silica GC columns and GC/MS systems
For use with SGE SilTite Ferrule Starter Kits
SilTite Nuts
  • Non-Swagelok™ nuts with two notches on outer surface
  • Inner taper of the nut corresponds to angle of ferrule, ensuring an even closure of the ferrule around the whole column to eliminate leaks
  • Ferrules are easily removed from nut while still attached to capillary column
  • Reusable
Specifications
For Use With (Equipment) Split/Splitless Injectors
Type Nut
Material Metal
For Use With (Application) Split/Splitless Injector
Quantity 5/Pk.