Description
- Next generation ferrule system for gas chromatography
- Eliminates leaks
- No contamination
- No need to retighten after temperature cycling
- Cleaner spectra for better library matches
Specifications
Specifications
| Type | Blank Ferrule |
| Material | SilTite |
| Quantity | 2/Pk. |