Learn More
Trajan™ SilTite™ FingerTite Female Nut
SDP
Catalog No. SG073636 Shop All SGE Inc. Products
Change view
Click to view available options
Packaging Quantity:
5/Pk.
Catalog No. Packaging Quantity
SG073636 5/Pk.
1 options
Catalog No. SG073636 Supplier Trajan™ Supplier No. 073636
SilTite FingerTite delivers an easy, leak-free installation for capillary columns without the use of any tools
  • Next generation ferrule system for gas chromatography
  • Eliminates leaks
  • No contamination
  • No need to retighten after temperature cycling
  • Cleaner spectra for better library matches
Specifications
Type Female Nut
Packaging Quantity 5/Pk.
For Use With Chromatography Ferrule
Material SilTite