Learn More
SGE™ SilTite™ FingerTite Ferrule System Starter Kits
SDP
Catalog No. SG073625 Shop All SGE Inc. Products
Change view
Click to view available options
For Use With:
Agilent Capillary/FID
Agilent INJ
Agilent INJ/FID
Generic
PerkinElmer
PerkinElmer INJ/FID
PerkinElmer INJ/MS
SGE SilTite
Shimadzu 17 A INJ/MS
Shimadzu 17AINJ/FID
Shimadzu 2010 INJ/FID
Shimadzu 2010 INJ/MS
Shimadzu Agilent INJ/FID
Shimadzu Agilent INJ/MS
Thermo INJ/DSQ/DSQII
Thermo INJ/FID
Thermo INJ/ISQ
Varian/Bruker INJ/FID
Varian/Bruker INJ/MS
21 product options available for selection
Product selection table with 21 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With
SG073625 Generic
SG073629 Shimadzu 17 A INJ/MS
SG073628 Shimadzu 17AINJ/FID
SG073610 Shimadzu Agilent INJ/FID
SG073621 Shimadzu 2010 INJ/MS
SG073612 Shimadzu Agilent INJ/MS
SG073620 Shimadzu 2010 INJ/FID
SG0736100 SGE SilTite
SG0736105 SGE SilTite
SG07361053 Agilent INJ/FID
SG07361054 Agilent INJ
SG073611 Agilent Capillary/FID
SG073614 Thermo INJ/FID
SG073615 Thermo INJ/ISQ
SG073616 Thermo INJ/DSQ/DSQII
SG073618 Varian/Bruker INJ/FID
SG073619 Varian/Bruker INJ/MS
SG07362053 Shimadzu 2010 INJ/FID
SG073622 PerkinElmer INJ/FID
SG07362254 PerkinElmer
SG073623 PerkinElmer INJ/MS
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 21 options available.
21 options
Catalog No. SG073625 Supplier SGE™ Supplier No. 073625
For gas chromatography systems
SilTite FingerTite is the next generation ferrule system for gas chromatography systems delivering an easy, leak free installation for capillary columns without the use of any tools. Each starter kit includes all the parts necessary to convert one GC system (one injector and one detector) to the SilTite FingerTite system. The system requires no permanent instrument modifications—switching back to conventional connections can be done quickly and easily.
  • Easy to use
  • Reliable connecton
  • Leak free every time
  • Removes contamination caused by Vespel™ or graphite ferrules
  • No need to re-tighten ferrules after temperature cycling
  • Reduce air background
  • NO TOOLS (FingerTite)
Specifications
Type FingerTite Ferrule Starter Kit
For Use With Generic