Learn More
Trajan™ Syringe with Valve
SDP
Catalog No. SG004279 Shop All SGE Inc. Products
Change view
Click to view available options
Volume (Metric):
50 μL
Catalog No. Volume (Metric)
SG004279 50 μL
1 options
Catalog No. SG004279 Supplier Trajan™ Supplier No. 004279
Ideal for sample storage and transportation or to pressurize sample prior to injection.
  • High quality solutions for liquid handling
  • Accurate and reproducible injections
  • Pre-fitted with a valve for sample pressurization prior to injection
Specifications
Material PTFE (Plunger)
Product Type Pre-fitted with Syringe Valves
Volume (Metric) 50 μL