Learn More
Trajan™ Thermo Scientific AS3000, AS2000 and AS200/800GC Autosampler Syringe
SDP
Catalog No. 03351108 Shop All SGE Inc. Products
Change view
Click to view available options
Quantity:
1/Pk.
6/Pk.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
03-351-108 1/Pk.
03-351-113 6/Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 03-351-108 Supplier Trajan™ Supplier No. 0029801
Fixed needle autosampler syringes. For reduced non specific sample interaction and increased analyte recovery
  • High quality solutions for liquid handling
  • Accurate and reproducible injections
  • Specifically designed for Thermo Scientific instruments
Specifications
Needle Gauge 26 G
Diameter (Metric) Outer 0.47 mm
Product Type AS3000, AS2000 and AS200/800GC Autosampler Syringe
Quantity 1/Pk.
Volume (Metric) 10 μL