Learn More
Trajan™ Thermo Scientific Instrument Ferrules
SDP
Catalog No. SG073452 Shop All SGE Inc. Products
Change view
Click to view available options
Diameter (Metric) Inside:
0.10 to 0.25 mm
0.32 mm
0.45 to 0.53 mm
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Diameter (Metric) Inside
SG073452 0.45 to 0.53 mm
SG073451 0.32 mm
SG073450 0.10 to 0.25 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. SG073452 Supplier Trajan™ Supplier No. 073452
Item Discontinued This item has been discontinued by the Fisher Scientific channel. Please to view product availability in your area.
SilTite FingerTite delivers an easy, leak-free installation for capillary columns without the use of any tools
  • Next generation ferrule system for gas chromatography
  • Eliminates leaks
  • No contamination
  • No need to retighten after temperature cycling
  • Cleaner spectra for better library matches
Specifications
Material SilTite Metal
Type GC Interface Connection
Quantity 10 ferrules, 2 nuts
Diameter (Metric) Inside 0.45 to 0.53 mm