Learn More
Trajan™ Thermo Scientific Instrument Ferrules
SDP
Catalog No. SG0726549 Shop All SGE Inc. Products
Change view
Click to view available options
Diameter (Metric) Inside:
0.10 to 0.25 mm
Catalog No. Diameter (Metric) Inside
SG0726549 0.10 to 0.25 mm
1 options
Catalog No. SG0726549 Supplier Trajan™ Supplier No. 0726549
Item Discontinued This item has been discontinued by the Fisher Scientific channel. Please to view product availability in your area.
SilTite FingerTite delivers an easy, leak-free installation for capillary columns without the use of any tools
  • Next generation ferrule system for gas chromatography
  • Eliminates leaks
  • No contamination
  • No need to retighten after temperature cycling
  • Cleaner spectra for better library matches
Specifications
Material Graphite, Vespel
Type Detector/Injectors
Diameter (Metric) Inside 0.10 to 0.25 mm