Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SGEF Antibody, DyLight 680, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP305912FR
Description
SGEF Polyclonal specifically detects SGEF in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.Specifications
SGEF | |
Polyclonal | |
Immunocytochemistry/Immunofluorescence | |
Rho guanine nucleotide exchange factor (GEF) 26 | |
Produced in rabbits immunized with E. coli-derived Human SGEF fragment. | |
0.1 mL | |
Primary | |
Human | |
Purified |
Immunofluorescence | |
DyLight 680 | |
50mM Sodium Borate | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
26084 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction