Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ SGEF Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5107064
Description
Sequence homology: Cow: 100%; Human: 100%; Mouse: 100%; Pig: 100%; Rabbit: 100%; Rat: 100%; Yeast: 90%.
Activates RhoG GTPase by promoting the exchange of GDP by GTP. Required for the formation of membrane ruffles during macropinocytosis. Required for the formation of cup-like structures during trans-endothelial migration of leukocytes. In case of Salmonella enterica infection, activated by SopB, which induces cytoskeleton rearrangements and promotes bacterial entry.
Specifications
SGEF | |
Polyclonal | |
Unconjugated | |
ARHGEF26 | |
4631416L12Rik; 8430436L14Rik; AI303572; ARHGEF26; AW491953; CSGEF; HMFN1864; RGD1562559; Rho guanine nucleotide exchange factor (GEF) 26; Rho guanine nucleotide exchange factor 26; SGEF; SH3 domain-containing guanine exchange factor; Src homology 3 domain-containing guanine nucleotide exchange factor; testicular tissue protein Li 171 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
26084 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide; pH 7.4 | |
Q96DR7 | |
ARHGEF26 | |
Synthetic peptide directed towards the N terminal region of human SGEF (aa 1-51). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction