Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SGEF Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA553626
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA553626 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA553626 Supplier Invitrogen™ Supplier No. PA553626
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: PEEDLTGLTA SPVPSPTANG LAANNDSPGS GSQSGRKAKD PERGLFPGPQ KSSSEQKLPL QRLPSQENEL LENPSVVLST NSPAALKVGK QQIIPKSLAS EIKISKSNNQ NV Highest antigen sequence identity to the following orthologs: Mouse - 70%, Rat - 71%.

Activates RhoG GTPase by promoting the exchange of GDP by GTP. Required for the formation of membrane ruffles during macropinocytosis. Required for the formation of cup-like structures during trans-endothelial migration of leukocytes. In case of Salmonella enterica infection, activated by SopB, which induces cytoskeleton rearrangements and promotes bacterial entry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SGEF
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ARHGEF26
Gene Accession No. Q96DR7
Gene Alias 4631416L12Rik; 8430436L14Rik; AI303572; ARHGEF26; AW491953; CSGEF; HMFN1864; RGD1562559; Rho guanine nucleotide exchange factor (GEF) 26; Rho guanine nucleotide exchange factor 26; SGEF; SH3 domain-containing guanine exchange factor; Src homology 3 domain-containing guanine nucleotide exchange factor; testicular tissue protein Li 171
Gene Symbols ARHGEF26
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human SGEF. Recombinant protein control fragment (Product #RP-90732).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 26084
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.