Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SGF29 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA562816
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA562816 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA562816 Supplier Invitrogen™ Supplier No. PA562816
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: RGVLMTLLQQ SAMTLPLWIG KPGDKPPPLC GAIPASGDYV ARPGDKVAAR VKAVDGDEQW ILAEVVSYSH ATN Highest antigen sequence identity to the following orthologs: Mouse - 97%, Rat - 97%.

CCDC101 is a subunit of 2 histone acetyltransferase complexes: the ADA2A -containing -TAF9 /PCAF acetylase (STAGA) complex. Both of these complexes contain either GCN5 or PCAF, which are paralogous acetyltransferases.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SGF29
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene SGF29
Gene Accession No. Q96ES7
Gene Alias CCDC101; coiled-coil domain containing 101; coiled-coil domain-containing protein 101; SAGA complex associated factor 29; SAGA complex-associated factor 29; SAGA-associated factor 29; SAGA-associated factor 29 homolog; SGF29; STAF36; TDRD29
Gene Symbols SGF29
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human SGF29. Recombinant protein control fragment (Product #RP-98592).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 112869
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.