Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SGIP1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA553681
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA553681 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA553681 Supplier Invitrogen™ Supplier No. PA553681
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: GTPPPLPPKN VPATPPRTGS PLTIGPGNDQ SATEVKIEKL PSINDLDSIF GPVLSPKSVA VNAEEKWVHF SDTSPEHVTP ELTPREKVVS PPATPDNPAD SP Highest antigen sequence identity to the following orthologs: Mouse - 86%, Rat - 87%.

SGIP1 functions as an endocytic protein that affects signaling by receptors in neuronal systems involved in energy homeostasis via its interaction with endophilins.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SGIP1
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene SGIP1
Gene Accession No. Q9BQI5
Gene Alias 3110007P09Rik; endophilin-3-interacting protein; gag; I79_011488; SGIP1; SGIP1alpha; SH3 domain GRB2 like endophilin interacting protein 1; SH3-containing GRB2-like protein 3-interacting protein 1; SH3-domain GRB2-like (endophilin) interacting protein 1; SH3-domain GRB2-like interacting protein 1 alpha
Gene Symbols SGIP1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human SGIP1. Recombinant protein control fragment (Product #RP-90737).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 84251
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.