Learn More
Promega SGK1 Kinase Enzyme System, 1mg
Catalog No. PRV2912 Shop All Promega Corporation Products
Encompass
Change view
Click to view available options
Quantity:
1 mg
Catalog No. Quantity
PRV2912 1 mg
1 options
Catalog No. PRV2912 Supplier Promega Supplier No. V2912
Add to Cart
Add to Cart
Request a Quote
Recombinant human SGK1 (amino acids 60-end) was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag. SGK1 is a member of the serum- and glucocorticoid-induced protein kinase family.
  • Easily Screen and Profile SGK1 Kinase Inhibitors
  • Includes kinase, substrate and reaction buffer
  • Use with ADP−Glo™ Assay for bioluminescent detection of kinase activity
Specifications
Quantity 1 mg
Content And Storage <-65°C