Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SGK1 Recombinant Rabbit Monoclonal Antibody (3P4N3)
GREENER_CHOICE

Catalog No. PIMA535420
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA535420 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA535420 Supplier Invitrogen™ Supplier No. MA535420
Add to Cart
Add to Cart

Rabbit Recombinant Monoclonal Antibody

Immunogen sequence: MTVKTEAAKG TLTYSRMRGM VAILIAFMKQ RRMGLNDFIQ KIANNSYACK HPEVQSILKI SQPQEPELMN ANPSPPPSPS QQINLGPSSN PHAKPSDFHF.

SGK (SGK1) is a serine/threonine kinase that may be a transcriptional target for p38/MAPK in hepatoma cells. SGK plays an important role in cellular stress response and cell survival. SGK1 is transcriptionally activated by numerous stimuli, including serum, glucocorticoids, osmotic stress, and hormones. It phosphorylates and negatively regulates pro-apoptotic FOXO3A. SGK1 is thought to play an essential function in nephropathy and cardiovascular development.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SGK1
Applications ELISA, Western Blot
Classification Recombinant Monoclonal
Clone 3P4N3
Concentration 1.06 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.05% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Gene SGK1
Gene Accession No. O00141, Q9WVC6
Gene Alias serine/threonine protein kinase SGK; serine/threonine-protein kinase Sgk1; serum and glucocorticoid-dependent protein kinase; serum/glucocorticoid regulated kinase 1; serum/glucocorticoid-regulated kinase 1; Sgk; SGK1; Sgk1 variant i3
Gene Symbols SGK1
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence within amino acids 1-100 of human SGK1 (O00141).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 20393, 6446
Target Species Human, Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.