Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ SGK1, Mouse, Unlabeled, Clone: 1G2.1,

Catalog No. MABS1476MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
Conjugate:
Unconjugated
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate
MABS1476MI 100 μg Unconjugated
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MABS1476MI Supplier MilliporeSigma™ Supplier No. MABS1476
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Serine/threonine-protein kinase Sgk1 (EC 2.7.11.1; UniProt O00141; also known as Serum/glucocorticoid-regulated kinase 1) is encoded by the SGK1 (also known as SGK) gene (Gene ID 6446) in human. SGK1 is a serine/threonine kinase that is induced in response to a variety of stimuli, including serum, osmotic shock, FSH, glucocorticoids, and mineralocorticoids. It is expressed in most tissues with highest levels observed in in the pancreas, followed by placenta, kidney, and lung. SGK1 is involved in the regulation of a wide variety of ion channels, membrane transporters, cellular enzymes, transcription factors, neuronal excitability, cell growth, proliferation, survival, migration and apoptosis. It is also reported to play an important role in cellular stress response. It is also involved in the regulation of renal sodium retention and potassium elimination. SGK1 activity is regulated by phosphorylation and is activated by phosphorylation on Ser422 by mTORC2. This phosphorylation transforms it into a substrate for PDPK1, which phosphorylates it on Thr256. Phosphorylation on Ser397 and Ser401 are also considered to be essential for its activity. It is negatively regulated by ubiquitination and proteasome degradation. High levels of expression of SGK1 gene may contribute to conditions such as hypertension and diabetic nephropathy. SGK1 has been reported to be up-regulated in non-small cell lung cancer (NSCLC) and its over-expression promotes the growth and migration of NSCLC cells via GSK-3 beta phosphorylation and activation of beta-catenin/TCF signaling. Ref.: Xiaobo, Y et al. (2016) Gene 576, 339-346.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SGK1
Applications Western Blot
Classification Monoclonal
Clone 1G2.1
Concentration Please refer to lot specific datasheet.
Conjugate Unconjugated
Formulation Purified mouse IgG1 in buffer containing 0.1M Tris-Glycine (pH 7.4), 150mM NaCl with 0.05% Sodium Azide.
Gene SGK1, SGK
Gene Accession No. O00141
Host Species Mouse
Immunogen GST-tagged recombinant human SGK1 fragment corresponding to the region N-terminal to the kinase domain.
Purification Method Protein G Purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Signaling
Primary or Secondary Primary
Gene ID (Entrez) NP_005618
Test Specificity Clone 1G2.1 was raised against SGK1 N-terminal fragment, reactivity towards human spliced isoforms 2-5 and murine spliced isoforms 2 and 3 are possible only if the epitope lies outside the first 25-amino acids of isoform 1.
Target Species Human
Content And Storage Stable for 1 year at 2°-8°C from date of receipt.
Form Purified
Isotype IgG1 κ
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.