Learn More
Invitrogen™ SGK1 Polyclonal Antibody

Description
This antibody is tested in Peptide ELISA: antibody detection limit dilution 1:4,000.
Specifications
Specifications
| Antigen | SGK1 |
| Applications | Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 |
| Gene | SGK1 |
| Gene Accession No. | O00141 |
| Gene Alias | serine/threonine protein kinase SGK; serine/threonine-protein kinase Sgk1; serum and glucocorticoid-dependent protein kinase; serum/glucocorticoid regulated kinase 1; serum/glucocorticoid-regulated kinase 1; Sgk; SGK1; Sgk1 variant i3 |
| Gene Symbols | SGK1 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.