Learn More
Invitrogen™ SGK1 Polyclonal Antibody

Description
Keep as concentrated solution. Predicted reactivity: Mouse (96%), Rat (96%), Zebrafish (88%), Xenopus laevis (91%), Pig (95%), Rabbit (95%), Chicken (94%), Chimpanzee (100%), Bovine (95%). Positive Control: mouse brain. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Specifications
Specifications
| Antigen | SGK1 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 1.38 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 20% glycerol and 0.01% thimerosal; pH 7 |
| Gene | SGK1 |
| Gene Accession No. | O00141, Q9WVC6 |
| Gene Alias | serine/threonine protein kinase SGK; serine/threonine-protein kinase Sgk1; serum and glucocorticoid-dependent protein kinase; serum/glucocorticoid regulated kinase 1; serum/glucocorticoid-regulated kinase 1; Sgk; SGK1; Sgk1 variant i3 |
| Gene Symbols | SGK1 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.