Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SGK1, Rabbit anti-Human, Polyclonal Antibody, Abnova™
Rabbit Polyclonal Antibody
Supplier: Abnova Corporation PAB31641
Specifications
SGK1 | |
Polyclonal | |
ELISA (1:10000) Western Blot (1:500-2000) The optimal working dilution should be determined by the end user. | |
serum/glucocorticoid regulated kinase 1 | |
SGK1 | |
A synthetic peptide corresponding to amino acids 10-90 of human SGK1 (phospho S78). | |
RUO | |
6446 | |
Human, Mouse, Rat | |
Antibody | |
IgG |
ELISA, Western Blot | |
Unconjugated | |
In PBS (50% glycerol, 0.5% BSA and 0.02% sodium azide). | |
SGK | |
Rabbit | |
100 μL | |
Primary | |
This antibody detects endogenous levels of SGK1 protein only when phosphorylated at S78. | |
Store at -20°C. Aliquot to avoid repeated freezing and thawing. | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction