Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Sgk110 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PA548684
Change view
Click to view available options
Quantity:
400 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PA548684 400 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PA548684 Supplier Invitrogen™ Supplier No. PA548684
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Sgk110 Polyclonal Antibody for Western Blot

Sgk110 is a protein coding gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Sgk110
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene Sbk3
Gene Accession No. P0C5K0
Gene Alias Gm1078; Sbk3; Sgk110; SH3 domain binding kinase family member 3; SH3 domain binding kinase family, member 3; SH3-binding domain kinase family member 3; Sugen kinase 110; uncharacterized serine/threonine-protein kinase SBK3
Gene Symbols Sbk3
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide between 332-360 amino acids from the C-terminal region of mouse Sgk110.
Purification Method Antigen affinity chromatography, Protein A
Quantity 400 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 381835
Target Species Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.