Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SGK2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA552824
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA552824 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA552824 Supplier Invitrogen™ Supplier No. PA552824
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: EVLRKEPYDR AVDWWCLGAV LYEMLHGLPP FYSQDVSQMY ENILHQPLQI PGGRTVAACD LLQSLLHKDQ RQRLGSKADF LEIKNHVFFS PINWDDLYHK RLTPPFNPNV TGPADLKHFD PEFTQEAVSK SIGCTPD Highest antigen sequence identity to the following orthologs: Mouse - 93%, Rat - 93%.

SGK2 is a serine/threonine protein kinase which is similar to serum- and glucocorticoid-induced protein kinase (SGK). Unlike SGK, SGK2 is not induced by serum or glucocorticoids. It is induced in response to signals that activate phosphatidylinositol 3-kinase, which is also true for SGK. Although little is known about the transcriptional regulation of SGK2, the protein has been shown to be phosphorylated and activated by 3 phosphoinositide dependent protein kinase 1 (PDK1), and to a lesser extent insulin like growth factor 1, in vitro.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SGK2
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene SGK2
Gene Accession No. Q8R4U9, Q9HBY8, Q9QZS5
Gene Alias AI098171; AW146006; dJ138B7.2; H-SGK2; Serine/threonine-protein kinase Sgk2; serum/glucocorticoid regulated kinase 2; serum/glucocorticoid-regulated kinase 2; Sgk2; SGK2, serine/threonine kinase 2; Sgkl
Gene Symbols SGK2
Host Species Rabbit
Immunogen Recombinant protein fragment of Human SGK2 (Product #RP-88957)
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10110, 171497, 27219
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.