Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ SGK2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5110756
Description
Immunogen sequence: EVLRKEPYDR AVDWWCLGAV LYEMLHGLPP FYSQDVSQMY ENILHQPLQI PGGRTVAACD LLQSLLHKDQ RQRLGSKADF LEIKNHVFFS PINWDDLYHK RLTPPFNPNV TGPADLKHFD PEFTQEAVSK SIGCTPD.
SGK2 is a serine/threonine protein kinase which is similar to serum- and glucocorticoid-induced protein kinase (SGK). Unlike SGK, SGK2 is not induced by serum or glucocorticoids. It is induced in response to signals that activate phosphatidylinositol 3-kinase, which is also true for SGK. Although little is known about the transcriptional regulation of SGK2, the protein has been shown to be phosphorylated and activated by 3 phosphoinositide dependent protein kinase 1 (PDK1), and to a lesser extent insulin like growth factor 1, in vitro.
Specifications
| SGK2 | |
| Polyclonal | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9HBY8 | |
| SGK2 | |
| Recombinant Protein corresonding to Human SGK2. Recombinant protein control fragment (Product #RP-88957). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
| Immunocytochemistry | |
| Unconjugated | |
| SGK2 | |
| AI098171; AW146006; dJ138B7.2; H-SGK2; Serine/threonine-protein kinase Sgk2; serum/glucocorticoid regulated kinase 2; serum/glucocorticoid-regulated kinase 2; Sgk2; SGK2, serine/threonine kinase 2; Sgkl | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 10110 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction