Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SGK223 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA567970
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA567970 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA567970 Supplier Invitrogen™ Supplier No. PA567970
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Predicted to react with Mouse and Rat samples.

PRAGMIN is also known as SGK223 (sugen kinase 223). The rat homolog was originally identified in a yeast two-hybrid screen for Rnd2 effector proteins expressed in neural cells. Rat Pragmin was found to stimulate RhoA activity for the regulation of neurite outgrowth.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SGK223
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene PRAG1
Gene Accession No. Q571I4, Q86YV5
Gene Alias 9830148H23; D8Ertd82e; DNA segment, Chr 8, ERATO Doi 82, expressed; homolog of rat pragma of Rnd2; Inactive tyrosine-protein kinase PRAG1; mFLJ00269; NACK; Notch activation complex kinase; PEAK1-related kinase-activating pseudokinase 1; Prag1; Pragma of Rnd2; pragmin; RGD1311793; Sgk223; Sugen kinase 223; tyrosine-protein kinase SgK223
Gene Symbols PRAG1
Host Species Rabbit
Immunogen Synthesized peptide derived from human SGK223.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 157285, 244418, 306506
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.