Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SGK223 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA552905
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA552905 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA552905 Supplier Invitrogen™ Supplier No. PA552905
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: TNGVAISDPS RCPQPAASSA SEQRRPRFQA GTWSRQCRIE EEEEVEQELL SHSWGRETKN GPTDHSNSTT WHRLHPTDGS SGQNSKVGTG MSKSASFAFE FPKD Highest antigen sequence identity to the following orthologs: Mouse - 74%, Rat - 75%.

PRAGMIN is also known as SGK223 (sugen kinase 223). The rat homolog was originally identified in a yeast two-hybrid screen for Rnd2 effector proteins expressed in neural cells. Rat Pragmin was found to stimulate RhoA activity for the regulation of neurite outgrowth.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SGK223
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.20 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene PRAG1
Gene Accession No. Q86YV5
Gene Alias 9830148H23; D8Ertd82e; DNA segment, Chr 8, ERATO Doi 82, expressed; homolog of rat pragma of Rnd2; Inactive tyrosine-protein kinase PRAG1; mFLJ00269; NACK; Notch activation complex kinase; PEAK1-related kinase-activating pseudokinase 1; Prag1; Pragma of Rnd2; pragmin; RGD1311793; Sgk223; Sugen kinase 223; tyrosine-protein kinase SgK223
Gene Symbols PRAG1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human SGK223. Recombinant protein control fragment (Product #RP-89816).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 157285
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.