Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SGK3 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP220337
Description
SGK3 Polyclonal specifically detects SGK3 in Human samples. It is validated for Western Blot.Specifications
SGK3 | |
Polyclonal | |
Unconjugated | |
PBS (pH 7.0), 1.0% BSA and 20% Glycerol with 0.01% Thimerosal | |
CISK, DKFZp781N0293, EC 2.7.11, EC 2.7.11.1, serine/threonine-protein kinase Sgk3, serum/glucocorticoid regulated kinase 3, serum/glucocorticoid regulated kinase family, member 3, serum/glucocorticoid regulated kinase-like, Serum/glucocorticoid-regulated kinase 3, Serum/glucocorticoid-regulated kinase-like, SGK2, SGKLcytokine-independent survival kinase | |
Rabbit | |
57 kDa | |
0.1 mL | |
GPCR, Protein Kinase | |
23678 | |
Human | |
IgG |
Western Blot | |
1mg/mL | |
Western Blot 1:500-1:3000 | |
Q96BR1 | |
SGK3 | |
Recombinant protein encompassing a sequence within the N-terminus region of human SGK3. The exact sequence is proprietary. | |
Affinity Purified | |
RUO | |
Primary | |
Human. Species reactivity based on sequence identity: Mouse (94%). | |
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction