Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SGK3 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$382.00 - $610.00
Specifications
Antigen | SGK3 |
---|---|
Dilution | Immunohistochemistry 1:20 - 1:50, Immunohistochemistry-Paraffin 1:20 - 1:50 |
Applications | Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Conjugate | Unconjugated |
Description
SGK3 Polyclonal specifically detects SGK3 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
SGK3 | |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Unconjugated | |
RUO | |
PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
23678 | |
This antibody was developed against Recombinant Protein corresponding to amino acids:DVAEMYDNILHKPLSLRPGVSLTAWSILEELLEKDRQNRLGAKEDFLEIQNHPFFESLSWADLVQKKIPPPFNPNVAGPDDIRNFDTAFTEETVPYSVCVSSDYSIVNASVLEADDAFVGFSY | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Immunohistochemistry 1:20 - 1:50, Immunohistochemistry-Paraffin 1:20 - 1:50 | |
Polyclonal | |
Rabbit | |
Human | |
CISK, DKFZp781N0293, EC 2.7.11, EC 2.7.11.1, serine/threonine-protein kinase Sgk3, serum/glucocorticoid regulated kinase 3, serum/glucocorticoid regulated kinase family, member 3, serum/glucocorticoid regulated kinase-like, Serum/glucocorticoid-regulated kinase 3, Serum/glucocorticoid-regulated kinase-like, SGK2, SGKLcytokine-independent survival kinase | |
SGK3 | |
IgG | |
Affinity Purified | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title